5,6-Dichloroindoline-2,3-dione 96%

Supplier: Ambeed
Warning

A219450-100MG A219450-250MG A219450-5G A219450-1G
77030-752EA 16.98 USD
77030-752 77030-754 77550-426 77593-428
5,6-Dichloroindoline-2,3-dione 96%
5,6-Dichloroindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₈H₃Cl₂NO₂
MW: 216.0200 g/mol
MDL Number: MFCD06797894
CAS Number: 1677-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR