(4-Ethoxy-2,6-dimethylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethyl-4-ethoxyphenylboronic acid

A219357-1G A219357-250MG A219357-5G
76999-866EA 33.95 USD
76999-866 76999-868 76999-870
(4-Ethoxy-2,6-dimethylphenyl)boronic acid 97%
(4-Ethoxy-2,6-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.03 g/mol
MDL Number: MFCD12026736
CAS Number: 1315342-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR