Fmoc-2-methyl-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(o-tolyl)propanoic acid

A219274-250MG A219274-5G A219274-10G A219274-1G
76999-862EA 11 USD
76999-862 76999-864 77671-956 77881-541
Fmoc-2-methyl-L-phenylalanine 95%
Fmoc-2-methyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MW: 401.47 g/mol
MDL Number: MFCD00671391
CAS Number: 211637-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR