Fmoc-N-methyl-D-valine 97%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-N-methyl-D-valine, Fmoc-D-N-Me-Val-OH

A219181-1G A219181-25G A219181-5G
77030-744EA 16 USD
77030-744 77030-746 77030-748
Fmoc-N-methyl-D-valine 97%
Fmoc-N-methyl-D-valine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄
MDL Number: MFCD00153396
CAS Number: 103478-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR