Ethyl-1H-indazole-6-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1H-indazole-6-carboxylate

A219092-100MG A219092-1G
77013-212EA 12.74 USD
77013-212 77013-214
Ethyl-1H-indazole-6-carboxylate 98%
Ethyl 1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2000 g/mol
MDL Number: MFCD07371613
CAS Number: 713-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR