D-(-)-N-Ethylglucamine 98%

Supplier: Ambeed
Warning

A219049-1000G A219049-100G A219049-25G A219049-500G
76999-854EA 99 USD
76999-854 76999-856 76999-858 76999-860
D-(-)-N-Ethylglucamine 98%
D-(-)-N-Ethylglucamine

Precaución: for Research Use Only

Formula: H₃CCH₂NHCH₂(CH(OH))₄CH₂OH
MDL Number: MFCD03789564
CAS Number: 14216-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR