7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine

A218977-250MG A218977-25G
77030-738EA 31.83 USD
77030-738 77568-120
7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine 98%
7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Precaución: For research use only.

Formula: C₆H₅BrN₄
MW: 213.0347 g/mol
MDL Number: MFCD11846606
CAS Number: 882521-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR