4,4'-(Hexane-1,6-diylbis(oxy))dibenzonitrile 95+%

Supplier: Ambeed
Warning

A218950-100G A218950-500G
76986-996EA 5 USD
76986-996 77612-134
4,4'-(Hexane-1,6-diylbis(oxy))dibenzonitrile 95+%
4,4'-(Hexane-1,6-diylbis(oxy))dibenzonitrile

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₂
MDL Number: MFCD01529716
CAS Number: 94291-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR