2-chloro-4-methoxy-1-nitrobenzene 98%

Supplier: Ambeed
Warning

A218895-100G A218895-1G A218895-250MG A218895-25G A218895-5G
76999-844EA 1680 USD
76999-844 76999-846 76999-848 76999-850 76999-852
2-chloro-4-methoxy-1-nitrobenzene 98%
2-chloro-4-methoxy-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD00156594
CAS Number: 28987-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR