4,6-Diphenylpyrimidin-2-amine 97%

Supplier: Ambeed
Warning

A218559-100MG A218559-1G A218559-250MG A218559-5G
76986-980EA 10 USD
76986-980 76986-982 76986-984 76986-986
4,6-Diphenylpyrimidin-2-amine 97%
4,6-Diphenylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃N₃
MDL Number: MFCD00234910
CAS Number: 40230-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR