Ticlopidine hydrochloride ≥99%, Ambeed.Inc

Supplier: Ambeed
Warning

A218498-1MG A218498-250MG A218498-25G A218498-5G A218498-1G
77013-200EA 5 USD
77013-200 77013-202 77013-204 77013-206 77683-630
Ticlopidine hydrochloride ≥99%, Ambeed.Inc
Ticlopidine hydrochloride

Precaución: For research use only.

Formula: C₁₄H₁₄ClNS
MDL Number: MFCD00079606
CAS Number: 53885-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR