N4-(3-Ethynylphenyl)-7-methoxyquinazoline-4,6-diamine 97%

Supplier: Ambeed
Warning

A218380-100MG A218380-250MG
77030-724EA 45.62 USD
77030-724 77030-726
N4-(3-Ethynylphenyl)-7-methoxyquinazoline-4,6-diamine 97%
N4-(3-Ethynylphenyl)-7-methoxyquinazoline-4,6-diamine

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₄O
MW: 290.31 g/mol
MDL Number: MFCD16877567
CAS Number: 1012057-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR