Dec-9-ynoic acid 97%

Supplier: Ambeed
Danger

Synonyms: 9-Decynoic acid

A218306-1G A218306-250MG A218306-100MG
77013-198EA 706.56 USD
77013-198 77612-130 77683-629
Dec-9-ynoic acid 97%
Dec-9-ynoic acid

Precaución: For research use only.

Formula: C₁₀H₁₆O₂
MW: 168.23 g/mol
MDL Number: MFCD08064029
CAS Number: 1642-49-5
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR