2-Chloro-5-methylthiazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A218299-100MG A218299-1G A218299-250MG
76999-828EA 29.71 USD
76999-828 76999-830 77671-948
2-Chloro-5-methylthiazole-4-carboxylic acid 98%
2-Chloro-5-methylthiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄ClNO₂S
MW: 177.609 g/mol
MDL Number: MFCD12404922
CAS Number: 1194374-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR