4-(Ethylamino)-2-(methylthio)pyrimidine-5-carbaldehyde 98+%

Supplier: Ambeed
Warning

A218248-100MG A218248-1G A218248-250MG
76986-966EA 16.98 USD
76986-966 76986-968 76986-970
4-(Ethylamino)-2-(methylthio)pyrimidine-5-carbaldehyde 98+%
4-(Ethylamino)-2-(methylthio)pyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₁N₃OS
MW: 197.25 g/mol
MDL Number: MFCD12911523
CAS Number: 185040-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR