1-Cyclopentyl-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A218213-100MG A218213-1G A218213-250MG
76986-960EA 36.08 USD
76986-960 76986-962 76986-964
1-Cyclopentyl-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
1-Cyclopentyl-3-iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₂IN₅
MW: 329.14 g/mol
MDL Number: MFCD26743438
CAS Number: 330794-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR