2,4-Dichloro-6-(4-methoxyphenyl)pyrimidine 97%

Supplier: Ambeed
Warning

A218195-100MG A218195-1G A218195-250MG A218195-5G
76986-952EA 40.32 USD
76986-952 76986-954 76986-956 76986-958
2,4-Dichloro-6-(4-methoxyphenyl)pyrimidine 97%
2,4-Dichloro-6-(4-methoxyphenyl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₈Cl₂N₂O
MW: 255.10 g/mol
MDL Number: MFCD18074237
CAS Number: 163263-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR