Ethyl 4-methyl-3-nitrobenzoate 95+%

Supplier: Ambeed
Warning

A218145-100G A218145-25G A218145-500G
77030-718EA 60.48 USD
77030-718 77030-720 77030-722
Ethyl 4-methyl-3-nitrobenzoate 95+%
Ethyl 4-methyl-3-nitrobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₄
MW: 209.199 g/mol
MDL Number: MFCD06204349
CAS Number: 19013-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR