2-Bromo-5-ethylpyridine 95%

Supplier: Ambeed
Warning

A217952-100MG A217952-5G A217952-1G A217952-250MG
77030-702EA 47.75 USD
77030-702 77030-704 77593-418 77667-822
2-Bromo-5-ethylpyridine 95%
2-Bromo-5-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.049 g/mol
MDL Number: MFCD11869552
CAS Number: 19842-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR