trans-Cyclopentane-1,3-diamine dihydrochloride 97%

Supplier: Ambeed
Warning

A217891-100MG A217891-1G A217891-250MG A217891-5G
77013-188EA 64 USD
77013-188 77013-190 77013-192 77013-194
trans-Cyclopentane-1,3-diamine dihydrochloride 97%
trans-Cyclopentane-1,3-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₄Cl₂N₂
MDL Number: MFCD27952970
CAS Number: 1799439-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR