6-Bromo-5-methoxy-2-pyridinecarbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-methoxypicolinaldehyde

A217869-100MG A217869-250MG
76999-824EA 81.69 USD
76999-824 76999-826
6-Bromo-5-methoxy-2-pyridinecarbaldehyde 97%
6-Bromo-5-methoxy-2-pyridinecarbaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.04 g/mol
MDL Number: MFCD13193086
CAS Number: 329217-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR