Ethyl-4-ethoxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-ethoxybenzoate, 4-Ethoxybenzoic acid ethyl ester

A217486-100G A217486-1G A217486-25G A217486-5G
76999-816EA 133.68 USD
76999-816 76999-818 76999-820 76999-822
Ethyl-4-ethoxybenzoate 98%
Ethyl-4-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
MDL Number: MFCD00009116
CAS Number: 23676-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR