4-(Diethylamino)salicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Diethylamino)-2-hydroxybenzaldehyde

A217387-100G A217387-500G A217387-25G
76999-812EA 19.1 USD
76999-812 76999-814 77087-968
4-(Diethylamino)salicylaldehyde 98%
4-(Diethylamino)salicylaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
MDL Number: MFCD00003326
CAS Number: 17754-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR