2-(3-(Aminomethyl)phenyl)ethanol 97%

Supplier: Ambeed
Warning

A217268-100MG A217268-250MG
76986-926EA 175.05 USD
76986-926 76986-928
2-(3-(Aminomethyl)phenyl)ethanol 97%
2-(3-(Aminomethyl)phenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.20 g/mol
MDL Number: MFCD20719358
CAS Number: 1379356-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR