3,3'-Oxybis(propan-1-ol) 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A217119-1G A217119-5G A217119-10G A217119-250MG A217119-25G
77013-170EA 40 USD
77013-170 77013-172 77683-626 77890-294 77890-295
3,3'-Oxybis(propan-1-ol) 98%, Ambeed.Inc
3,3'-Oxybis(propan-1-ol)

Precaución: For research use only.

Formula: C₆H₁₄O₃
MDL Number: MFCD00051023
CAS Number: 2396-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR