tert-Butyl N-[(5-chloro-2-pyridyl)methyl]carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl ((5-chloropyridin-2-yl)methyl)carbamate

A217034-1G A217034-250MG
76986-920EA 39 USD
76986-920 76986-922
tert-Butyl N-[(5-chloro-2-pyridyl)methyl]carbamate 97%
tert-Butyl N-[(5-chloro-2-pyridyl)methyl]carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O₂
MDL Number: MFCD19982297
CAS Number: 67938-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR