(S)-2-Amino-2-(4-fluorophenyl)ethanol hydrochloride 95%

Supplier: Ambeed
Warning

A217023-100MG A217023-1G A217023-250MG
76986-914EA 35.01 USD
76986-914 76986-916 76986-918
(S)-2-Amino-2-(4-fluorophenyl)ethanol hydrochloride 95%
(S)-2-Amino-2-(4-fluorophenyl)ethanol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClFNO
MW: 191.63 g/mol
MDL Number: MFCD12758115
CAS Number: 1269773-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR