5,6-Dimethyl-1H-benzimidazole-2-thiol 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dimethyl-2-mercapto-1H-benzimidazole, 5,6-Dimethyl-1H-benzo[d]imidazole-2(3H)-thione

A216923-1G A216923-250MG A216923-5G A216923-25G
77013-156EA 178 USD
77013-156 77013-158 77013-160 77890-290
5,6-Dimethyl-1H-benzimidazole-2-thiol 95%
5,6-Dimethyl-1H-benzimidazole-2-thiol

Precaución: for Research Use Only

Formula: C₉H₁₀N₂S
MW: 178.26 g/mol
MDL Number: MFCD00519115
CAS Number: 3287-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR