4-Bromo-2,6-dichloropyridin-3-amine 97%

Supplier: Ambeed
Warning

A216829-1G A216829-250MG
76986-910EA 45.62 USD
76986-910 76986-912
4-Bromo-2,6-dichloropyridin-3-amine 97%
4-Bromo-2,6-dichloropyridin-3-amine

Precaución: for Research Use Only

Formula: C₅H₃BrCl₂N₂
MW: 241.90 g/mol
MDL Number: MFCD12546348
CAS Number: 1121586-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR