3-Acetylphenyl ethyl(methyl)carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Rivastigmine Related Compound D

A216758-10G A216758-1G A216758-5G A216758-250MG A216758-100MG
77013-144EA 502 USD
77013-144 77013-146 77013-148 77593-412 77612-126
3-Acetylphenyl ethyl(methyl)carbamate 98%
3-Acetylphenyl ethyl(methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD20257239
CAS Number: 855300-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR