Methyl 2-amino-5-bromo-4-methoxybenzoate 95%

Supplier: Ambeed
Warning

A216747-100MG A216747-1G A216747-250MG A216747-25G A216747-5G
77013-134EA 100.79 USD
77013-134 77013-136 77013-138 77013-140 77013-142
Methyl 2-amino-5-bromo-4-methoxybenzoate 95%
Methyl 2-amino-5-bromo-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₃
MW: 260.08 g/mol
MDL Number: MFCD21868787
CAS Number: 169044-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR