6-Bromo-3-methoxypicolinic acid 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A216593-1G A216593-250MG A216593-5G
76986-906EA 210.06 USD
76986-906 77612-122 77683-623
6-Bromo-3-methoxypicolinic acid 95%, Ambeed.Inc
6-Bromo-3-methoxypicolinic acid

Precaución: For research use only.

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD11846420
CAS Number: 1256810-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR