3-Chloro-2-fluoroanisole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-fluoro-3-methoxybenzene

A216563-10G A216563-5G
76999-804EA 47.75 USD
76999-804 76999-806
3-Chloro-2-fluoroanisole 95%
3-Chloro-2-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD01631570
CAS Number: 261762-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR