(3-Fluoropyridin-4-yl)methanamine 97%

Supplier: Ambeed
Warning

A216509-250MG A216509-5G A216509-1G
77030-686EA 26 USD
77030-686 77030-688 77872-103
(3-Fluoropyridin-4-yl)methanamine 97%
(3-Fluoropyridin-4-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MDL Number: MFCD11100727
CAS Number: 870063-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR