N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-S-(triphenylmethyl)-L-cysteine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-N-Me-Cys(Trt)-OH, Fmoc-MeCys(Trt)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-(tritylthio)propanoic acid, Fmoc-N-methyl-L-cysteine(Trityl)
A216450-1G
A216450-250MG
A216450-5G
A216450-25G
77013-116EA
93.36
USD
77013-116
77013-118
77550-400
77683-622
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-S-(triphenylmethyl)-L-cysteine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-S-(triphenylmethyl)-L-cysteine
Precaución: For research use only.
Formula:
C₃₈H₃₃NO₄S MW: 599.75 g/mol |
MDL Number:
MFCD11973909 CAS Number: 944797-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...