Norfloxacin nicotinate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid nicotinic acid salt

A216241-100G A216241-10G A216241-1G A216241-25G A216241-5G
76986-894EA 65.78 USD
76986-894 76986-896 76986-898 76986-900 76986-902
Norfloxacin nicotinate 95%
Norfloxacin nicotinate

Precaución: for Research Use Only

Formula: C₁₆H₁₈FN₃O₃·C₆H₅NO₂
MW: 319.3 · 123.1 g/mol
MDL Number: MFCD00873666
CAS Number: 118803-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR