Ethyl-(S)-4-Chloro-3-hydroxybutyrate 97%
Supplier: Ambeed
|
Danger
|
A216236-100G
A216236-10G
A216236-500G
A216236-25G
77030-678EA
305.54
USD
77030-678
77030-680
77030-682
77030-684
77087-830
Ethyl-(S)-4-Chloro-3-hydroxybutyrate 97%
Ethyl-(S)-4-Chloro-3-hydroxybutyrate
Precaución: for Research Use Only
Formula:
C₆H₁₁ClO₃ |
MDL Number:
MFCD00211241 CAS Number: 86728-85-0 UN: 1760 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...