Methyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate 98%

Supplier: Ambeed
Warning

A216199-100MG A216199-250MG A216199-1G
76986-890EA 23.34 USD
76986-890 76986-892 77683-616
Methyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate 98%
Methyl 8-methyl-1,4-dioxaspiro[4.5]decane-8-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₈O₄
MW: 214.25 g/mol
MDL Number: MFCD22681691
CAS Number: 87787-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR