6-Methoxypyridine-3,4-diamine 97%

Supplier: Ambeed
Warning

A216169-100MG A216169-250MG A216169-1G
76999-796EA 29 USD
76999-796 76999-798 77881-534
6-Methoxypyridine-3,4-diamine 97%
6-Methoxypyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₆H₉N₃O
MDL Number: MFCD09744015
CAS Number: 127356-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR