(R)-5-Bromo-3-[(1-methyl-2-pyrrolidinyl)methyl]-1H-indole 95%

Supplier: Ambeed
Warning

A215953-1G A215953-25G A215953-5G
76999-782EA 19.1 USD
76999-782 76999-784 76999-786
(R)-5-Bromo-3-[(1-methyl-2-pyrrolidinyl)methyl]-1H-indole 95%
(R)-5-Bromo-3-[(1-methyl-2-pyrrolidinyl)methyl]-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₇BrN₂
MW: 293.20 g/mol
MDL Number: MFCD00912970
CAS Number: 143322-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR