Ethylsulfamoyl chloride 95%

Supplier: Ambeed
Danger

A215939-1G A215939-250MG A215939-100MG
77013-104EA 184 USD
77013-104 77013-106 77890-281
Ethylsulfamoyl chloride 95%
Ethylsulfamoyl chloride

Precaución: for Research Use Only

Formula: C₂H₆ClNO₂S
MDL Number: MFCD12401754
CAS Number: 16548-07-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR