3-Bromo-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidine 95%
Supplier: Ambeed
|
Warning
|
A215781-1G
A215781-250MG
77013-100EA
172
USD
77013-100
77013-102
3-Bromo-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidine 95%
3-Bromo-6-(methylthio)-1H-pyrazolo[3,4-d]pyrimidine
Precaución: for Research Use Only
Formula:
C₆H₅BrN₄S |
MDL Number:
MFCD22415216 CAS Number: 1306829-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...