5-Formylpyridine-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinaldehyde

A215551-1G A215551-250MG A215551-5G A215551-25G
77013-094EA 35 USD
77013-094 77013-096 77013-098 77890-279
5-Formylpyridine-3-boronic acid pinacol ester 98%
5-Formylpyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BNO₃
MW: 233.074 g/mol
MDL Number: MFCD04038759
CAS Number: 848093-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR