2-Cyanophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Cyanobenzeneboronic acid pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyanophenylboronic acid, pinacol ester

A215214-100G A215214-10G A215214-1G A215214-250MG A215214-25G A215214-5G
77013-082EA 140 USD
77013-082 77013-084 77013-086 77013-088 77013-090 77013-092
2-Cyanophenylboronic acid pinacol ester 97%
2-Cyanophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₂
MDL Number: MFCD04038747
CAS Number: 214360-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR