4,4'-Dibromo-2,2'-diiodobiphenyl 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Dibromo-2,2'-diiodo-1,1'-biphenyl

A215135-100MG A215135-250MG A215135-25G A215135-5G A215135-1G
76986-872EA 13 USD
76986-872 76986-874 76986-876 76986-878 77568-078
4,4'-Dibromo-2,2'-diiodobiphenyl 95%
4,4'-Dibromo-2,2'-diiodobiphenyl

Precaución: For research use only.

Formula: C₁₂H₆Br₂I₂
MW: 563.80 g/mol
MDL Number: MFCD14582983
CAS Number: 852138-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR