1-Bromo-3-chloro-2-iodobenzene 97%

Supplier: Ambeed
Warning

A215038-10G A215038-1G A215038-250MG A215038-25G A215038-5G A215038-100G A215038-500G
77030-624EA 21 USD
77030-624 77030-626 77030-628 77030-630 77030-632 77667-802 77872-099
1-Bromo-3-chloro-2-iodobenzene 97%
1-Bromo-3-chloro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClI
MDL Number: MFCD09907935
CAS Number: 450412-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR