2-sec-Butyl-3-methoxypyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(sec-Butyl)-3-methoxypyrazine

A214956-10G A214956-1G A214956-25G A214956-5G
76999-744EA 66 USD
76999-744 76999-746 76999-748 76999-750
2-sec-Butyl-3-methoxypyrazine 97%
2-sec-Butyl-3-methoxypyrazine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O
MDL Number: MFCD00006136
CAS Number: 24168-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR