4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Boc-amino)cyclohexanecarboxylic Acid, 4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid (cis- and trans- mixture), 4-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid

A214749-100G A214749-10G A214749-1G A214749-25G A214749-5G
77013-066EA 359 USD
77013-066 77013-068 77013-070 77013-072 77612-110
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 97%
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture)

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.30 g/mol
MDL Number: MFCD03453262
CAS Number: 130309-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR