5-Nitro-6-methyluracil 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-5-nitrouracil, 2,4-Dihydroxy-6-methyl-5-nitropyrimidine

A214683-1000G A214683-100G A214683-10G A214683-1G A214683-25G A214683-500G A214683-5G
77013-048EA 742.63 USD
77013-048 77013-050 77013-052 77013-054 77013-056 77013-058 77013-060
5-Nitro-6-methyluracil 97%
5-Nitro-6-methyluracil

Precaución: for Research Use Only

Formula: C5H5N3O4
MW: 171.11 g/mol
MDL Number: MFCD00047362
CAS Number: 16632-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR