11-Maleimidoundecanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 11-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)undecanoic acid

A214596-1G A214596-25G A214596-100G A214596-5G
77030-616EA 10.61 USD
77030-616 77600-728 77671-908 77671-910
11-Maleimidoundecanoic acid 97%
11-Maleimidoundecanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₃NO₄
MW: 281.352 g/mol
MDL Number: MFCD00941279
CAS Number: 57079-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR